Assay Details
Gene Symbol: Stag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016964.2 | NP_058660.2 | 9 - 10 | 1142 | 63 | |
GenBank mRNA | AJ005678.1 | - | 9 - 10 | 1100 | 63 | |
AK145898.1 | - | 9 - 10 | 1128 | 63 | ||
AK158240.1 | - | 9 - 10 | 1038 | 63 | ||
BC060184.1 | - | 9 - 10 | 1663 | 63 | ||
BC139010.1 | - | 9 - 10 | 1072 | 8862032 | 63 | |
BC139011.1 | - | 9 - 10 | 1072 | 8862033 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
