Assay Details
| Gene Symbol: Klhl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177755.3 | NP_808423.1 | 1 - 2 | 212 | 73 | |
| GenBank mRNA | AK033353.1 | - | 1 - 2 | 213 | 73 | |
| BC132483.1 | - | 1 - 2 | 114 | 40130760 | 73 | |
| BC145690.1 | - | 1 - 2 | 114 | 40130993 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033353.1, BC132483.1, BC145690.1, NM_177755.3 transcripts.
Set Membership: |
|

Klhl38
Genomic Map