Assay Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026346.3 | NP_080622.1 | 5 - 6 | 749 | 95 | |
| GenBank mRNA | AF441120.1 | - | 5 - 6 | 795 | 95 | |
| AK014612.1 | - | 5 - 6 | 702 | 95 | ||
| AK014793.1 | - | 5 - 6 | 731 | 95 | ||
| BC027211.1 | - | 5 - 6 | 683 | 3981597 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo32
Genomic Map