Assay Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026346.3 | NP_080622.1 | 6 - 7 | 931 | 59 | |
| GenBank mRNA | AF441120.1 | - | 6 - 7 | 977 | 59 | |
| AK014612.1 | - | 6 - 7 | 884 | 59 | ||
| AK014793.1 | - | 6 - 7 | 913 | 59 | ||
| BC027211.1 | - | 6 - 7 | 865 | 3981597 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo32
Genomic Map