Assay Details
| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005847.2 | NP_001005847.1 | 3 - 4 | 504 | 129 | |
| NM_001205054.1 | NP_001191983.1 | 3 - 4 | 504 | 129 | ||
| GenBank mRNA | AA396103.1 | - | 3 - 4 | 348 | 129 | |
| BC048230.1 | - | 3 - 4 | 1385 | 129 | ||
| BC132227.1 | - | 3 - 4 | 479 | 40130504 | 129 | |
| BC132229.1 | - | 3 - 4 | 479 | 40130506 | 129 | |
| BC145297.1 | - | 3 - 4 | 479 | 9053851 | 129 | |
| BQ571087.1 | - | 3 - 4 | 396 | 129 | ||
| S81393.1 | - | 3 - 4 | 421 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aga
Genomic Map