Assay Details
| Gene Symbol: Aga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005847.2 | NP_001005847.1 | 4 - 5 | 605 | 78 | |
| NM_001205054.1 | NP_001191983.1 | 4 - 5 | 605 | 78 | ||
| GenBank mRNA | AA396103.1 | - | 4 - 5 | 449 | 78 | |
| BC048230.1 | - | 4 - 5 | 1486 | 78 | ||
| BC132227.1 | - | 4 - 5 | 580 | 40130504 | 78 | |
| BC132229.1 | - | 4 - 5 | 580 | 40130506 | 78 | |
| BC145297.1 | - | 4 - 5 | 580 | 9053851 | 78 | |
| BQ571087.1 | - | 4 - 5 | 497 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aga
Genomic Map