Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197991.2 | NP_932108.2 | 6 - 7 | 823 | 73 | |
GenBank mRNA | AK150499.1 | - | 6 - 7 | 728 | 73 | |
AK160274.1 | - | 6 - 7 | 740 | 73 | ||
AK169010.1 | - | 6 - 7 | 774 | 73 | ||
AK171657.1 | - | 6 - 7 | 737 | 73 | ||
AK183758.1 | - | 2 - 3 | 118 | 73 | ||
AY761098.1 | - | 6 - 7 | 716 | 73 | ||
BC006065.1 | - | 4 - 5 | 460 | 73 | ||
BC020179.1 | - | 6 - 7 | 718 | 4913500 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
