Assay Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013470.2 | NP_038498.2 | 1 - 2 | 181 | 89 | |
| GenBank mRNA | AJ001633.1 | - | 1 - 2 | 164 | 88 | |
| AK090055.1 | - | 1 - 2 | 199 | 89 | ||
| AK150922.1 | - | 1 - 2 | 182 | 89 | ||
| AK150951.1 | - | 1 - 2 | 149 | 89 | ||
| AK151771.1 | - | 1 - 2 | 141 | 89 | ||
| AK152837.1 | - | 1 - 2 | 108 | 89 | ||
| AK152863.1 | - | 1 - 2 | 169 | 89 | ||
| AK168396.1 | - | 1 - 2 | 77 | 89 | ||
| AK168507.1 | - | 1 - 2 | 109 | 89 | ||
| AK169113.1 | - | 1 - 2 | 57 | 89 | ||
| AK169423.1 | - | 1 - 2 | 63 | 89 | ||
| BC090634.1 | - | 1 - 2 | 175 | 30942562 | 89 | |
| BC141167.1 | - | 1 - 2 | 135 | 9055779 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ001633.1, AK090055.1, AK150922.1, AK150951.1, AK151771.1, AK152837.1, AK152863.1, AK168396.1, AK168507.1, AK169113.1, AK169423.1, BC090634.1, BC141167.1, NM_013470.2 transcripts.
Set Membership: |
|

Anxa3
Genomic Map