Assay Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090055.1 | - | 3 - 4 | 340 | 66 | |
| BC141167.1 | - | 3 - 4 | 276 | 9055779 | 66 | |
| CT010280.1 | - | 1 - 2 | 105 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa3
Genomic Map