Assay Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013470.2 | NP_038498.2 | 7 - 8 | 697 | 68 | |
| GenBank mRNA | AJ001633.1 | - | 7 - 8 | 680 | 68 | |
| AK013850.1 | - | 5 - 6 | 450 | 68 | ||
| AK090055.1 | - | 7 - 8 | 715 | 68 | ||
| AK150922.1 | - | 7 - 8 | 698 | 68 | ||
| AK150951.1 | - | 7 - 8 | 665 | 68 | ||
| AK151771.1 | - | 7 - 8 | 657 | 68 | ||
| AK152837.1 | - | 7 - 8 | 624 | 68 | ||
| AK152863.1 | - | 7 - 8 | 685 | 68 | ||
| AK166389.1 | - | 7 - 8 | 669 | 68 | ||
| AK168396.1 | - | 7 - 8 | 593 | 68 | ||
| AK168507.1 | - | 7 - 8 | 625 | 68 | ||
| AK169113.1 | - | 7 - 8 | 573 | 68 | ||
| BC090634.1 | - | 7 - 8 | 691 | 30942562 | 68 | |
| BC141167.1 | - | 7 - 8 | 651 | 9055779 | 68 | |
| CT010280.1 | - | 5 - 6 | 480 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa3
Genomic Map