Assay Details
| Gene Symbol: Mrpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039084.1 | NP_001034173.1 | 1 - 2 | 83 | 74 | |
| NM_053158.3 | NP_444388.2 | 1 - 2 | 83 | 74 | ||
| GenBank mRNA | AK010343.1 | - | 1 - 2 | 57 | 75 | |
| AK015476.1 | - | 1 - 2 | 43 | 74 | ||
| AK017938.1 | - | 1 - 2 | 55 | 74 | ||
| AK083978.1 | - | 1 - 2 | 58 | 74 | ||
| AK142249.1 | - | 1 - 2 | 58 | 74 | ||
| AK172544.1 | - | 1 - 2 | 48 | 74 | ||
| BC061042.1 | - | 1 - 2 | 47 | 6706599 | 74 | |
| BX527348.1 | - | 2 - 3 | 191 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010343.1, AK015476.1, BC061042.1 transcripts.
Set Membership: |
|

Mrpl1
Genomic Map