Assay Details
| Gene Symbol: Mrpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039084.1 | NP_001034173.1 | 5 - 6 | 628 | 70 | |
| NM_053158.3 | NP_444388.2 | 5 - 6 | 628 | 70 | ||
| GenBank mRNA | AB049632.1 | - | 4 - 5 | 584 | 70 | |
| AK010343.1 | - | 5 - 6 | 602 | 70 | ||
| AK015476.1 | - | 5 - 6 | 588 | 70 | ||
| AK017938.1 | - | 5 - 6 | 600 | 70 | ||
| AK083978.1 | - | 5 - 6 | 604 | 70 | ||
| AK142249.1 | - | 5 - 6 | 603 | 70 | ||
| AK172544.1 | - | 5 - 6 | 593 | 70 | ||
| BC061042.1 | - | 5 - 6 | 592 | 6706599 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl1
Genomic Map