Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 2 - 3 | 820 | 69 | |
| GenBank mRNA | AF516680.1 | - | 2 - 3 | 784 | 69 | |
| AK154473.1 | - | 2 - 3 | 821 | 69 | ||
| AK172982.1 | - | 1 - 2 | 198 | 69 | ||
| BC053101.1 | - | 2 - 3 | 796 | 6405486 | 69 | |
More Information
Set Membership: |
|

Pom121
Genomic Map