Assay Details
| Gene Symbol: Pom121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148932.2 | NP_683734.2 | 8 - 9 | 1528 | 92 | |
| GenBank mRNA | AK154473.1 | - | 8 - 9 | 1529 | 92 | |
| AK172982.1 | - | 7 - 8 | 906 | 92 | ||
| BC053101.1 | - | 8 - 9 | 1504 | 6405486 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pom121
Genomic Map