Assay Details
| Gene Symbol: Upk3bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027158.1 | NP_081434.1 | 2 - 3 | 376 | 112 | |
| GenBank mRNA | AK009780.1 | - | 2 - 3 | 355 | 112 | |
| BC099515.1 | - | 2 - 3 | 274 | 112 | ||
| BC115563.1 | - | 2 - 3 | 376 | 112 | ||
| BC115564.1 | - | 2 - 3 | 376 | 40102008 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upk3bl
Genomic Map