Assay Details
| Gene Symbol: Upk3bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027158.1 | NP_081434.1 | 4 - 5 | 666 | 58 | |
| GenBank mRNA | AK009780.1 | - | 4 - 5 | 645 | 58 | |
| BC099515.1 | - | 4 - 5 | 564 | 58 | ||
| BC115563.1 | - | 4 - 5 | 666 | 58 | ||
| BC115564.1 | - | 4 - 5 | 666 | 40102008 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057296 | BC099515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057296 | BX638176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057296 | NM_027158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057296 | AK009780.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29404 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7235 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110445 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057296 | BC099515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057296 | BX638176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057296 | NM_027158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057296 | AK009780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01229567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110445 | - | - |
| Whole Mouse Genome G4122A | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345340 | BX638176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345340 | NM_027158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345340 | AK009780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345344 | BC099515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345344 | BC115563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345344 | BC115564.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345344 | NM_027158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P345344 | AK009780.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107767 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04212023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04364480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04785635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04970039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04721025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04734992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04134596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04673480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04363371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24119223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526454 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429844_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526452 | - | - |
| Mouse Genome 430 2.0 Array | 1429844_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429844_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69665_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69665_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500014422.mm.1 | TC0500001596.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500014422.mm.1 | NM_027158.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500014422.mm.1 | NM_027158.1 | Refseq |
| Mouse Expression Set 430 | 1429844_at | - | - |
| Mouse Expression Set 430 | 1429844_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429844_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429844_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 458524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684465 | - | - |

Upk3bl
Genomic Map