Assay Details
| Gene Symbol: Upk3bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009780.1 | - | 5 - 6 | 768 | 76 | |
| BC099515.1 | - | 5 - 6 | 687 | 76 | ||
| BC115564.1 | - | 5 - 6 | 789 | 40102008 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upk3bl
Genomic Map