Assay Details
| Gene Symbol: Vwce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027913.1 | NP_082189.1 | 14 - 15 | 1998 | 62 | |
| GenBank mRNA | AK153937.1 | - | 14 - 15 | 1778 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vwce
Genomic Map