Assay Details
| Gene Symbol: Prpf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253843.1 | NP_001240772.1 | 1 - 2 | 236 | 92 | |
| NM_134129.4 | NP_598890.1 | 1 - 2 | 236 | 92 | ||
| GenBank mRNA | AF251503.1 | - | 1 - 2 | 201 | 92 | |
| AK047693.1 | - | 1 - 2 | 54 | 92 | ||
| AK088769.1 | - | 1 - 2 | 221 | 92 | ||
| AK164681.1 | - | 1 - 2 | 224 | 92 | ||
| BC004070.1 | - | 1 - 2 | 175 | 3591050 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF251503.1,AK047693.1,AK088769.1,AK164681.1,BC004070.1,NM_001253843.1,NM_134129.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prpf19
Genomic Map