Assay Details
| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146094.2 | NP_666206.1 | 6 - 7 | 979 | 71 | |
| GenBank mRNA | AB072976.1 | - | 6 - 7 | 894 | 71 | |
| AF061504.1 | - | 6 - 7 | 881 | 71 | ||
| AK033308.1 | - | 4 - 5 | 336 | 71 | ||
| AK083959.1 | - | 6 - 7 | 841 | 71 | ||
| AK154367.1 | - | 6 - 7 | 929 | 71 | ||
| AK193280.1 | - | 2 - 3 | 128 | 71 | ||
| AK198806.1 | - | 2 - 3 | 128 | 71 | ||
| AK213965.1 | - | 2 - 3 | 129 | 71 | ||
| BC022139.1 | - | 6 - 7 | 924 | 5100199 | 71 | |
| BC026831.1 | - | 6 - 7 | 946 | 4239695 | 71 | |
| BC026848.1 | - | 6 - 7 | 919 | 4196030 | 71 | |
| BC063053.1 | - | 6 - 7 | 839 | 5696722 | 71 | |
| KF987079.1 | - | 6 - 7 | 863 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fads1
Genomic Map