Assay Details
| Gene Symbol: 1700012I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045140.1 | 4 - 5 | 481 | 68 | ||
| GenBank mRNA | AK005914.1 | - | 4 - 5 | 482 | 68 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BY706053.1 | - | 485 | Chr. - - [-] on Build | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST3 beta-galactoside alpha-2,3-sialyltransferase 1 | St3gal1 |
5330418N22Rik, AI467004, ST3GalI, Siat4, Siat4a, St3gal-1 |
-74.8kb | Anti-sense | Chr. 15 - 67102875 - 67176910 on NCBI Build GRCm38 |
| Downstream | zinc finger and AT hook domain containing | Zfat |
Gm922, Zfat1, Zfp406 |
798.6kb | Anti-sense | Chr. 15 - 68083714 - 68259265 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700012I11Rik
Genomic Map