Assay Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008318.3 | NP_032344.2 | 1 - 2 | 65 | 110 | |
| GenBank mRNA | AK132371.1 | - | 1 - 2 | 64 | 110 | |
| AK162640.1 | - | 1 - 2 | 48 | 110 | ||
| BB618282.1 | - | 1 - 2 | 182 | 110 | ||
| BC045143.1 | - | 1 - 2 | 50 | 6391115 | 110 | |
| L20232.1 | - | 1 - 2 | 47 | 110 | ||
| L23801.1 | - | 1 - 2 | 66 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132371.1, AK162640.1, BC045143.1, L20232.1, L23801.1, NM_008318.3 transcripts.
Set Membership: |
|

Ibsp
Genomic Map