Assay Details
| Gene Symbol: Ibsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008318.3 | NP_032344.2 | 6 - 7 | 506 | 77 | |
| GenBank mRNA | AK132371.1 | - | 6 - 7 | 505 | 77 | |
| BC045143.1 | - | 6 - 7 | 491 | 6391115 | 77 | |
| L20232.1 | - | 6 - 7 | 488 | 77 | ||
| L23801.1 | - | 6 - 7 | 507 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986813 | BB373876.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986813 | BC045143.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986813 | NM_008318.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986813 | AK132371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | AK016300.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | AK018311.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | AK032228.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | AK042771.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | BC089589.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | NM_029545.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802475 | XM_915203.2 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139240 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126637 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115897 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075485 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986813 | BB373876.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986813 | BC045143.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986813 | NM_008318.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986813 | AK132371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486923 | BC045143.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486923 | L23801.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486923 | NM_008318.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486923 | AK132371.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486923 | AK162640.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23951435 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126637 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115897 | - | - |
| Whole Mouse Genome G4122A | A_51_P486923 | BC045143.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486923 | L23801.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486923 | NM_008318.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486923 | AK132371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P486923 | AK162640.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114037 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122351 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126637 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115897 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04669281 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04669287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04669270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04669279 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04669286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23951540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04758746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04969725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04338529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04669277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04338531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04331222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04331217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04669280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04380696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04338527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04758728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04450591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23951414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523708 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523705 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417485_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417484_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523701 | - | - |
| Mouse Genome 430 2.0 Array | 1417485_at | - | - |
| Mouse Genome 430 2.0 Array | 1417484_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417485_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417484_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15891_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15891_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500008624.mm.1 | TC0500001023.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500008624.mm.1 | NM_008318.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500008624.mm.1 | NM_008318.3 | Refseq |
| Mouse Expression Set 430 | 1417485_PM_at | - | - |
| Mouse Expression Set 430 | 1417484_at | - | - |
| Mouse Expression Set 430 | 1417484_PM_at | - | - |
| Mouse Expression Set 430 | 1417485_at | - | - |
| Murine Genome U74v2 Set | 100476_at | - | - |
| Murine Genome U74v2 Set | 100476_PM_at | - | - |
| Murine Genome U74v2 Set | 97572_PM_at | - | - |
| Murine Genome U74v2 Set | 97572_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417484_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417485_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417484_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417485_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 658958 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2060048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770088 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770088 | - | - |
| mouse-6 v1.1 (Illumina) | scl27512.8.140_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27512.8.140_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3063340 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3141106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3141106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3063340 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770088 | - | - |

Ibsp
Genomic Map