Assay Details
| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128212.1 | - | 3 - 4 | 283 | 92 | |
| AF128213.1 | - | 3 - 4 | 283 | 92 | ||
| BC116851.1 | - | 3 - 4 | 287 | 92 | ||
| BC116855.1 | - | 3 - 4 | 287 | 92 | ||
| M86672.1 | - | 3 - 4 | 385 | 92 | ||
More Information
Set Membership: |
|

Il12a
Genomic Map