Assay Details
| Gene Symbol: Il12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF128210.1 | - | 4 - 5 | 446 | 81 | |
| AF128211.1 | - | 4 - 5 | 446 | 81 | ||
| AF128212.1 | - | 5 - 6 | 373 | 81 | ||
| AF128213.1 | - | 5 - 6 | 373 | 81 | ||
| BC116851.1 | - | 5 - 6 | 377 | 81 | ||
| BC116855.1 | - | 5 - 6 | 377 | 81 | ||
| M86672.1 | - | 5 - 6 | 475 | 81 | ||
More Information
Set Membership: |
|

Il12a
Genomic Map