Assay Details
| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016689.2 | NP_057898.2 | 1 - 2 | 185 | 138 | |
| GenBank mRNA | AB019039.1 | - | 1 - 2 | 102 | 138 | |
| AK145678.1 | - | 1 - 2 | 186 | 138 | ||
| AK160929.1 | - | 1 - 2 | 187 | 138 | ||
| BC027400.1 | - | 1 - 2 | 182 | 4951027 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aqp3
Genomic Map