Assay Details
| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016689.2 | NP_057898.2 | 2 - 3 | 306 | 76 | |
| GenBank mRNA | AB019039.1 | - | 2 - 3 | 223 | 76 | |
| AK145678.1 | - | 2 - 3 | 307 | 76 | ||
| AK160929.1 | - | 2 - 3 | 308 | 76 | ||
| BC027400.1 | - | 2 - 3 | 303 | 4951027 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aqp3
Genomic Map