Assay Details
| Gene Symbol: Nfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290448.1 | NP_001277377.1 | 6 - 7 | 1558 | 95 | |
| NM_001290449.1 | NP_001277378.1 | 6 - 7 | 1558 | 95 | ||
| NM_023739.3 | NP_076228.2 | 6 - 7 | 1521 | 95 | ||
| GenBank mRNA | AF223576.1 | - | 6 - 7 | 1485 | 95 | |
| AK005038.2 | - | 6 - 7 | 1493 | 95 | ||
| AK033850.1 | - | 6 - 7 | 1452 | 95 | ||
| AK050749.1 | - | 6 - 7 | 1495 | 95 | ||
| AK144647.1 | - | 6 - 7 | 1507 | 95 | ||
| AK146108.1 | - | 6 - 7 | 1475 | 95 | ||
| AK155387.1 | - | 6 - 7 | 1473 | 95 | ||
| AK160013.1 | - | 6 - 7 | 1491 | 95 | ||
| AK178098.1 | - | 4 - 5 | 339 | 95 | ||
| BC053709.1 | - | 6 - 7 | 1456 | 30052746 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nfx1
Genomic Map