Assay Details
| Gene Symbol: Nfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290448.1 | NP_001277377.1 | 7 - 8 | 1702 | 76 | |
| NM_001290449.1 | NP_001277378.1 | 7 - 8 | 1702 | 76 | ||
| NM_023739.3 | NP_076228.2 | 7 - 8 | 1665 | 76 | ||
| GenBank mRNA | AF223576.1 | - | 7 - 8 | 1629 | 76 | |
| AK005038.2 | - | 7 - 8 | 1637 | 76 | ||
| AK033850.1 | - | 7 - 8 | 1596 | 76 | ||
| AK050749.1 | - | 7 - 8 | 1639 | 76 | ||
| AK144647.1 | - | 7 - 8 | 1651 | 76 | ||
| AK146108.1 | - | 7 - 8 | 1619 | 76 | ||
| AK155387.1 | - | 7 - 8 | 1617 | 76 | ||
| AK160013.1 | - | 7 - 8 | 1635 | 76 | ||
| BC053709.1 | - | 7 - 8 | 1600 | 30052746 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nfx1
Genomic Map