Assay Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007473.4 | NP_031499.1 | 1 - 2 | 263 | 64 | |
| GenBank mRNA | AB010100.1 | - | 1 - 2 | 154 | 64 | |
| AB056091.1 | - | 1 - 2 | 97 | 64 | ||
| AK076642.1 | - | 1 - 2 | 231 | 64 | ||
| AK131744.1 | - | 1 - 2 | 231 | 64 | ||
| BU945845.1 | - | 1 - 2 | 266 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB010100.1, AB056091.1, AK076642.1, AK131744.1, NM_007473.4 transcripts.
Set Membership: |
|

Aqp7
Genomic Map