Assay Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007473.4 | NP_031499.1 | 5 - 6 | 807 | 61 | |
| GenBank mRNA | AB010100.1 | - | 5 - 6 | 698 | 61 | |
| AB056091.1 | - | 5 - 6 | 641 | 61 | ||
| AK076642.1 | - | 5 - 6 | 775 | 61 | ||
| AK131744.1 | - | 5 - 6 | 775 | 61 | ||
| BC022223.1 | - | 3 - 4 | 437 | 4242417 | 61 | |
More Information
Set Membership: |
|

Aqp7
Genomic Map