Assay Details
| Gene Symbol: Dydc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027717.1 | NP_081993.1 | 1 - 2 | 77 | 71 | |
| GenBank mRNA | AK016963.1 | - | 1 - 2 | 212 | 71 | |
| BC113750.1 | - | 1 - 2 | 77 | 40049377 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016963.1, BC113750.1, NM_027717.1 transcripts.
Set Membership: |
|

Dydc2
Genomic Map