Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 1 - 2 | 223 | 117 | |
| GenBank mRNA | AK047411.1 | - | 1 - 2 | 92 | 117 | |
| AK053420.1 | - | 1 - 2 | 197 | 117 | ||
| AK084627.1 | - | 1 - 2 | 204 | 117 | ||
| AK140534.1 | - | 1 - 2 | 182 | 117 | ||
| AK160956.1 | - | 1 - 2 | 242 | 117 | ||
| AK165012.1 | - | 1 - 2 | 212 | 117 | ||
| BC057962.1 | - | 1 - 2 | 177 | 5353891 | 117 | |
| BY743247.1 | - | 1 - 2 | 224 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047411.1 transcripts.
Set Membership: |
|

Guf1
Genomic Map