Assay Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172711.2 | NP_766299.1 | 6 - 7 | 732 | 96 | |
| GenBank mRNA | AK032597.1 | - | 5 - 6 | 1740 | 96 | |
| AK047411.1 | - | 6 - 7 | 601 | 96 | ||
| AK053420.1 | - | 6 - 7 | 706 | 96 | ||
| AK084627.1 | - | 6 - 7 | 713 | 96 | ||
| AK140534.1 | - | 6 - 7 | 691 | 96 | ||
| AK160956.1 | - | 6 - 7 | 751 | 96 | ||
| AK165012.1 | - | 6 - 7 | 721 | 96 | ||
| BC057962.1 | - | 6 - 7 | 686 | 5353891 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Guf1
Genomic Map