Assay Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009758.4 | NP_033888.2 | 4 - 5 | 637 | 91 | |
| GenBank mRNA | AK132051.1 | - | 4 - 5 | 650 | 91 | |
| AY365062.1 | - | 2 - 3 | 231 | 91 | ||
| BC042611.1 | - | 3 - 4 | 427 | 5364272 | 91 | |
| BU840836.1 | - | 4 - 5 | 654 | 91 | ||
| D16250.1 | - | 2 - 3 | 372 | 91 | ||
| U04672.1 | - | 2 - 3 | 241 | 91 | ||
| Z23154.1 | - | 2 - 3 | 231 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr1a
Genomic Map