Assay Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009758.4 | NP_033888.2 | 8 - 9 | 1082 | 66 | |
| GenBank mRNA | AK132051.1 | - | 8 - 9 | 1095 | 66 | |
| AY365062.1 | - | 6 - 7 | 676 | 66 | ||
| BC042611.1 | - | 7 - 8 | 872 | 5364272 | 66 | |
| D16250.1 | - | 6 - 7 | 817 | 66 | ||
| U04672.1 | - | 6 - 7 | 686 | 66 | ||
| Z23154.1 | - | 6 - 7 | 676 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr1a
Genomic Map