Assay Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009758.4 | NP_033888.2 | 10 - 11 | 1570 | 128 | |
| GenBank mRNA | AK132051.1 | - | 10 - 11 | 1583 | 128 | |
| AK143804.1 | - | 1 - 2 | 197 | 128 | ||
| AY365062.1 | - | 8 - 9 | 1164 | 128 | ||
| BC042611.1 | - | 9 - 10 | 1360 | 5364272 | 128 | |
| D16250.1 | - | 8 - 9 | 1305 | 128 | ||
| U04672.1 | - | 8 - 9 | 1174 | 128 | ||
| Z23154.1 | - | 8 - 9 | 1164 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr1a
Genomic Map