Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 16 - 17 | 1682 | 98 | |
| GenBank mRNA | AF263460.1 | - | 14 - 15 | 1236 | 98 | |
| AK028885.1 | - | 16 - 17 | 1649 | 98 | ||
| AK033990.1 | - | 707 | 98 | |||
| AK035541.1 | - | 16 - 17 | 1669 | 98 | ||
| AK038559.1 | - | 15 - 16 | 1588 | 98 | ||
| AK082853.1 | - | 16 - 17 | 1684 | 98 | ||
| AK143277.1 | - | 16 - 17 | 1683 | 98 | ||
| AK149549.1 | - | 8 - 9 | 4133 | 98 | ||
| AY682914.1 | - | 17 - 18 | 1675 | 98 | ||
| BC023833.1 | - | 8 - 9 | 786 | 98 | ||
| BC024849.1 | - | 3 - 4 | 415 | 98 | ||
| BC026565.1 | - | 12 - 13 | 1100 | 98 | ||
| BC027806.1 | - | 16 - 17 | 1444 | 98 | ||
| BC031705.1 | - | 13 - 14 | 1196 | 98 | ||
| BC055773.1 | - | 13 - 14 | 1135 | 98 | ||
| BC078440.1 | - | 14 - 15 | 1259 | 98 | ||
| BC141008.1 | - | 17 - 18 | 1648 | 9055620 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map