Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 1 - 2 | 243 | 161 | |
| GenBank mRNA | AK028885.1 | - | 1 - 2 | 210 | 161 | |
| AK035541.1 | - | 1 - 2 | 230 | 161 | ||
| AK038559.1 | - | 1 - 2 | 208 | 161 | ||
| AK082853.1 | - | 1 - 2 | 245 | 162 | ||
| AK143277.1 | - | 1 - 2 | 244 | 161 | ||
| AY682914.1 | - | 1 - 2 | 236 | 161 | ||
| BC141008.1 | - | 1 - 2 | 209 | 9055620 | 161 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028885.1, AK035541.1, AY682914.1, BC141008.1, NM_178651.3 transcripts.
Set Membership: |
|

Slc30a9
Genomic Map