Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 7 - 8 | 808 | 106 | |
| GenBank mRNA | AF263460.1 | - | 5 - 6 | 362 | 106 | |
| AK028885.1 | - | 7 - 8 | 775 | 106 | ||
| AK035541.1 | - | 7 - 8 | 795 | 106 | ||
| AK038559.1 | - | 6 - 7 | 714 | 106 | ||
| AK082853.1 | - | 7 - 8 | 810 | 106 | ||
| AK143277.1 | - | 7 - 8 | 809 | 106 | ||
| AY682914.1 | - | 8 - 9 | 801 | 106 | ||
| BC026565.1 | - | 3 - 4 | 226 | 106 | ||
| BC027806.1 | - | 7 - 8 | 570 | 106 | ||
| BC031705.1 | - | 4 - 5 | 322 | 106 | ||
| BC055773.1 | - | 4 - 5 | 261 | 106 | ||
| BC078440.1 | - | 5 - 6 | 385 | 106 | ||
| BC141008.1 | - | 8 - 9 | 774 | 9055620 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map