Assay Details
| Gene Symbol: N4bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024917.1 | NP_001020088.1 | 16 - 17 | 5070 | 154 | |
| GenBank mRNA | AK136182.1 | - | 6 - 7 | 2217 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

N4bp2
Genomic Map