Assay Details
Gene Symbol: Nkain3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290410.1 | NP_001277339.1 | 2 - 3 | 548 | 75 | |
NM_172987.2 | NP_766575.2 | 2 - 3 | 548 | 75 | ||
GenBank mRNA | AK049776.1 | - | 2 - 3 | 748 | 75 | |
AK053823.1 | - | 2 - 3 | 433 | 75 | ||
AK141456.1 | - | 2 - 3 | 448 | 75 | ||
BC075683.1 | - | 2 - 3 | 548 | 30613986 | 75 | |
EF058052.1 | - | 2 - 3 | 195 | 75 | ||
EF058053.1 | - | 2 - 3 | 195 | 75 | ||
EF058054.1 | - | 2 - 3 | 195 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
