Assay Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161420.1 | NP_001154892.1 | 8 - 9 | 1781 | 66 | |
| NM_013703.2 | NP_038731.2 | 8 - 9 | 1781 | 66 | ||
| GenBank mRNA | AK076172.1 | - | 8 - 9 | 1786 | 66 | |
| AK136838.1 | - | 8 - 9 | 1601 | 66 | ||
| BC013622.1 | - | 8 - 9 | 1217 | 3968213 | 66 | |
| L33417.1 | - | 8 - 9 | 1205 | 66 | ||
| U06670.1 | - | 8 - 9 | 1367 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vldlr
Genomic Map