Assay Details
| Gene Symbol: Arhgef7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113517.1 | NP_001106989.1 | 16 - 17 | 2094 | 117 | |
| NM_001113518.1 | NP_001106990.1 | 15 - 16 | 1883 | 117 | ||
| NM_017402.4 | NP_059098.2 | 15 - 16 | 1883 | 117 | ||
| GenBank mRNA | AF247654.1 | - | 15 - 16 | 1779 | 117 | |
| AF343877.1 | - | 15 - 16 | 1763 | 117 | ||
| AK052545.1 | - | 15 - 16 | 1854 | 117 | ||
| AK129064.1 | - | 16 - 17 | 2030 | 117 | ||
| AK139156.1 | - | 16 - 17 | 2023 | 117 | ||
| BC044838.1 | - | 15 - 16 | 1868 | 3499604 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef7
Genomic Map