Assay Details
| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024248.1 | NP_077210.1 | 10 - 11 | 1106 | 64 | |
| GenBank mRNA | AK009937.1 | - | 10 - 11 | 1068 | 64 | |
| AK038968.1 | - | 10 - 11 | 1116 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cars2
Genomic Map