Assay Details
| Gene Symbol: Cars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024248.1 | NP_077210.1 | 3 - 4 | 443 | 105 | |
| GenBank mRNA | AK009937.1 | - | 3 - 4 | 405 | 105 | |
| AK038968.1 | - | 3 - 4 | 456 | 105 | ||
| AK041962.1 | - | 3 - 4 | 417 | 105 | ||
| AK054486.1 | - | 3 - 4 | 440 | 105 | ||
| CX204497.1 | - | 3 - 4 | 444 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cars2
Genomic Map