Assay Details
| Gene Symbol: Gyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294138.1 | NP_001281067.1 | 13 - 14 | 1354 | 68 | |
| NM_001294140.1 | NP_001281069.1 | 13 - 14 | 1288 | 68 | ||
| NM_008194.3 | NP_032220.1 | 12 - 13 | 1270 | 68 | ||
| NM_212444.2 | NP_997609.1 | 12 - 13 | 1270 | 68 | ||
| GenBank mRNA | AK044308.1 | - | 13 - 14 | 1126 | 68 | |
| AK088373.1 | - | 12 - 13 | 1089 | 68 | ||
| AK169424.1 | - | 13 - 14 | 1160 | 68 | ||
| AK169525.1 | - | 12 - 13 | 1271 | 68 | ||
| BC003767.1 | - | 12 - 13 | 1073 | 3593896 | 68 | |
| BQ555961.1 | - | 2 - 3 | 135 | 68 | ||
| CB950214.1 | - | 7 - 8 | 452 | 68 | ||
| CK789590.1 | - | 7 - 8 | 420 | 68 | ||
| U48403.1 | - | 12 - 13 | 1047 | 68 | ||
More Information
Set Membership: |
|

Gyk
Genomic Map