Assay Details
| Gene Symbol: Tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025729.4 | NP_080005.2 | 8 - 9 | 2297 | 79 | |
| GenBank mRNA | AK220185.1 | - | 8 - 9 | 2297 | 79 | |
| BC139024.1 | - | 7 - 8 | 2154 | 8862046 | 79 | |
| BC139025.1 | - | 7 - 8 | 2154 | 8862047 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tab3
Genomic Map