Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281969.1 | NP_001268898.1 | 3 - 4 | 353 | 158 | |
| NM_010298.6 | NP_034428.2 | 3 - 4 | 403 | 158 | ||
| GenBank mRNA | AK083251.1 | - | 2 - 3 | 257 | 158 | |
| AM420316.1 | - | 2 - 3 | 236 | 158 | ||
| BC027094.1 | - | 3 - 4 | 357 | 158 | ||
| BC037605.1 | - | 3 - 4 | 371 | 5361798 | 158 | |
| U09399.1 | - | 3 - 4 | 312 | 158 | ||
| X81202.1 | - | 2 - 3 | 195 | 158 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281969.1 transcripts.
Set Membership: |
|

Glrb
Genomic Map