Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281969.1 | NP_001268898.1 | 4 - 5 | 411 | 113 | |
| NM_010298.6 | NP_034428.2 | 4 - 5 | 461 | 113 | ||
| GenBank mRNA | AK083251.1 | - | 3 - 4 | 315 | 113 | |
| AM420316.1 | - | 3 - 4 | 294 | 113 | ||
| BC027094.1 | - | 4 - 5 | 415 | 113 | ||
| BC037605.1 | - | 4 - 5 | 429 | 5361798 | 113 | |
| U09399.1 | - | 4 - 5 | 370 | 113 | ||
| X81202.1 | - | 3 - 4 | 253 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001281969.1 transcripts.
Set Membership: |
|

Glrb
Genomic Map